missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(4-Vinylcyclohexyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5564006

 View more versions of this product

Catalog No. AAH5564006



Specifications

Specifications

trans-4-(4-Vinylcyclohexyl)benzonitrile
5g
C15H17N
4-4-ethenylcyclohexyl benzonitrile
C=CC1CCC(CC1)C2=CC=C(C=C2)C#N
211.308
211.31
57°C
96184-42-8
MFCD11053490
JHXJAWXZLWDDED-UHFFFAOYSA-N
4-(4-ethenylcyclohexyl)benzonitrile
18724353
97%

Chemical Identifiers

96184-42-8
211.308
JHXJAWXZLWDDED-UHFFFAOYSA-N
18724353
C=CC1CCC(CC1)C2=CC=C(C=C2)C#N
C15H17N
MFCD11053490
4-4-ethenylcyclohexyl benzonitrile
4-(4-ethenylcyclohexyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

Documents