missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(4-n-Pentylcyclohexyl)phenyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5532406

 View more versions of this product

Catalog No. AAH5532406



Specifications

Specifications

trans-4-(4-n-Pentylcyclohexyl)phenyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate
5g
C33H52O2
1alpha-4beta-1alpha-4beta-propylcyclohexyl cyclohexanecarboxylic acid 4-1alpha-4beta-pentylcyclohexyl phenyl ester
CCCCCC1CCC(CC1)C2=CC=C(C=C2)OC(=O)C3CCC(CC3)C4CCC(CC4)CCC
480.777
452.72
213°C to 223°C
131790-57-3
MFCD11053459
HNSOCAVSNSWNJJ-UHFFFAOYSA-N
[4-(4-pentylcyclohexyl)phenyl] 4-(4-propylcyclohexyl)cyclohexane-1-carboxylate
14993612
97%

Chemical Identifiers

131790-57-3
480.777
HNSOCAVSNSWNJJ-UHFFFAOYSA-N
14993612
CCCCCC1CCC(CC1)C2=CC=C(C=C2)OC(=O)C3CCC(CC3)C4CCC(CC4)CCC
C33H52O2
MFCD11053459
1alpha-4beta-1alpha-4beta-propylcyclohexyl cyclohexanecarboxylic acid 4-1alpha-4beta-pentylcyclohexyl phenyl ester
[4-(4-pentylcyclohexyl)phenyl] 4-(4-propylcyclohexyl)cyclohexane-1-carboxylate
Documents