missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(4-n-Pentylcyclohexyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5561203

 View more versions of this product

Catalog No. AAH5561203



Specifications

Specifications

trans-4-(4-n-Pentylcyclohexyl)benzonitrile
30°C to 55°C
61204-01-1
MFCD00075548
FURZYCFZFBYJBT-UHFFFAOYSA-N
4-(4-pentylcyclohexyl)benzonitrile
109063
97%
113°C (235°F)
1g
C18H25N
1-trans-4-amylcyclohexyl-4-cyanobenzene
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C#N
255.405
255.4

Chemical Identifiers

61204-01-1
255.405
FURZYCFZFBYJBT-UHFFFAOYSA-N
109063
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C#N
C18H25N
MFCD00075548
1-trans-4-amylcyclohexyl-4-cyanobenzene
4-(4-pentylcyclohexyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 262-656-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents