missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(4-n-Heptylcyclohexyl)benzonitrile, 97%

$148.49 - $589.80

Chemical Identifiers

CAS 61204-03-3
Molecular Formula C20H29N
Molecular Weight (g/mol) 283.459
MDL Number MFCD01941020
InChI Key NSGMZTNTQKRAFA-UHFFFAOYSA-N
Synonym 4-1s,4r-4-heptylcyclohexyl benzonitrile
PubChem CID 109064
IUPAC Name 4-(4-heptylcyclohexyl)benzonitrile
SMILES CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5542903 View Documents Alfa Aesar™
H5542903
1g Each for $148.49
 
AAH5542906 View Documents Alfa Aesar™
H5542906
5g Each for $589.80
 
Description

Description

Applications
It is used as intermediates of liquid crystals.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
Specifications

Chemical Identifiers

61204-03-3
283.459
NSGMZTNTQKRAFA-UHFFFAOYSA-N
109064
CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)C#N
C20H29N
MFCD01941020
4-1s,4r-4-heptylcyclohexyl benzonitrile
4-(4-heptylcyclohexyl)benzonitrile

Specifications

33°C
61204-03-3
MFCD01941020
Slightly soluble in water.
CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)C#N
283.459
283.46
trans-4-(4-n-Heptylcyclohexyl)benzonitrile
1g
C20H29N
4-1s,4r-4-heptylcyclohexyl benzonitrile
NSGMZTNTQKRAFA-UHFFFAOYSA-N
4-(4-heptylcyclohexyl)benzonitrile
109064
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 262-658-7

TSCA : Yes

Recommended Storage : Ambient temperatures