missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(4-n-Heptylcyclohexyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5542906

 View more versions of this product

Catalog No. AAH5542906



Description

Description

Applications
It is used as intermediates of liquid crystals.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
Specifications

Specifications

trans-4-(4-n-Heptylcyclohexyl)benzonitrile
5g
C20H29N
4-1s,4r-4-heptylcyclohexyl benzonitrile, 4-4-heptylcyclohexyl benzonitrile, 4-trans-4-heptylcyclohexyl benzonitrile, benzonitrile, 4-trans-4-heptylcyclohexyl, nsgmztntqkrafa-uhfffaoysa, p-4-heptylcyclohexyl benzonitrile, pch-7, trans-4-4-heptylcyclohexyl benzonitrile, trans-4-4-heptylcyclohexylbenzonitrile, trans-4-heptyl 4-cyanophenyl cyclohexane
NSGMZTNTQKRAFA-UHFFFAOYSA-N
4-(4-heptylcyclohexyl)benzonitrile
109064
97%
33°C
61204-03-3
MFCD01941020
Slightly soluble in water.
CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)C#N
283.459
283.46

Chemical Identifiers

61204-03-3
283.459
NSGMZTNTQKRAFA-UHFFFAOYSA-N
109064
CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)C#N
C20H29N
MFCD01941020
4-1s,4r-4-heptylcyclohexyl benzonitrile, 4-4-heptylcyclohexyl benzonitrile, 4-trans-4-heptylcyclohexyl benzonitrile, benzonitrile, 4-trans-4-heptylcyclohexyl, nsgmztntqkrafa-uhfffaoysa, p-4-heptylcyclohexyl benzonitrile, pch-7, trans-4-4-heptylcyclohexyl benzonitrile, trans-4-4-heptylcyclohexylbenzonitrile, trans-4-heptyl 4-cyanophenyl cyclohexane
4-(4-heptylcyclohexyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 262-658-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents