missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(4-n-Heptylcyclohexyl)benzoic acid, 98%

Manufacturer:  Alfa Aesar™ H6400506

 View more versions of this product

Catalog No. AAH6400506



Specifications

Specifications

trans-4-(4-n-Heptylcyclohexyl)benzoic acid
101532-38-1
MFCD09838992
1-bromo-4-1s,4r-4-heptylcyclohexyl benzene, 1-bromo-4-4-heptylcyclohexyl benzene, 1-bromo-4-trans-4-heptylcyclohexyl benzene
CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)Br
337.345
302.46
98%
5g
C19H29Br
4748130
OKIVMWBXCXBOGL-UHFFFAOYSA-N
1-bromo-4-(4-heptylcyclohexyl)benzene
15135337
Solid

Chemical Identifiers

101532-38-1
337.345
OKIVMWBXCXBOGL-UHFFFAOYSA-N
15135337
CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)Br
C19H29Br
MFCD09838992
1-bromo-4-1s,4r-4-heptylcyclohexyl benzene, 1-bromo-4-4-heptylcyclohexyl benzene, 1-bromo-4-trans-4-heptylcyclohexyl benzene
1-bromo-4-(4-heptylcyclohexyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents