Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4,4'-Diphenylstilbene, 99%

$56.50 - $141.03

Chemical Identifiers

Chemical Name or Material trans-4,4'-Diphenylstilbene
CAS 2039-68-1
Molecular Formula C26H20
Formula Weight 332.45
MDL Number MFCD00003063
Beilstein 2053491
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0783703

  SDS
alfa aesar™
L0783703
1g Each for $141.03

AAL07837MD

  SDS
alfa aesar™
L07837MD
250mg Each for $56.50
Description & Specifications

Specifications

Formula Weight 332.45
Chemical Name or Material trans-4,4'-Diphenylstilbene
MDL Number MFCD00003063
CAS 2039-68-1
Assay 99%
Melting Point 308° to 310°C
Quantity 1g
Molecular Formula C26H20
Beilstein 2053491