missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-4-(2-Nitrovinyl)benzeneboronic acid, 97%

$83.42 - $255.99

Chemical Identifiers

CAS 216394-04-6
Molecular Formula C8H8BNO4
Molecular Weight (g/mol) 192.965
MDL Number MFCD01075748
InChI Key GMEGTAODWQPCOD-AATRIKPKSA-N
Synonym e-4-2-nitrovinyl phenyl boronic acid, 4-2-nitrovinyl phenylboronic acid, 4-e-2-nitrovinyl phenylboronic acid, trans-4-beta-nitrovinyl benzeneboronic acid, 4-e-2-nitrovinyl benzeneboronic acid, 4-e-2-nitroethenyl phenyl boronic acid, trans-4-2-nitrovinyl benzeneboronic acid, 4-e-2-nitroethenyl phenyl boronic acid, 4-e-2-nitroethenyl phenylboronic acid, 4-e-2-nitroethenyl phenyl boronic acid
PubChem CID 5708381
IUPAC Name [4-[(E)-2-nitroethenyl]phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C=C[N+](=O)[O-])(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1700403 View Documents Alfa Aesar™
L1700403
1g Each for $255.99
 
AAL17004MD View Documents Alfa Aesar™
L17004MD
250mg Each for $83.42
 
Specifications

Chemical Identifiers

216394-04-6
192.965
GMEGTAODWQPCOD-AATRIKPKSA-N
5708381
B(C1=CC=C(C=C1)C=C[N+](=O)[O-])(O)O
C8H8BNO4
MFCD01075748
e-4-2-nitrovinyl phenyl boronic acid, 4-2-nitrovinyl phenylboronic acid, 4-e-2-nitrovinyl phenylboronic acid, trans-4-beta-nitrovinyl benzeneboronic acid, 4-e-2-nitrovinyl benzeneboronic acid, 4-e-2-nitroethenyl phenyl boronic acid, trans-4-2-nitrovinyl benzeneboronic acid, 4-e-2-nitroethenyl phenyl boronic acid, 4-e-2-nitroethenyl phenylboronic acid, 4-e-2-nitroethenyl phenyl boronic acid
[4-[(E)-2-nitroethenyl]phenyl]boronic acid

Specifications

289°C to 291°C
216394-04-6
MFCD01075748
e-4-2-nitrovinyl phenyl boronic acid, 4-2-nitrovinyl phenylboronic acid, 4-e-2-nitrovinyl phenylboronic acid, trans-4-beta-nitrovinyl benzeneboronic acid, 4-e-2-nitrovinyl benzeneboronic acid, 4-e-2-nitroethenyl phenyl boronic acid, trans-4-2-nitrovinyl benzeneboronic acid, 4-e-2-nitroethenyl phenyl boronic acid, 4-e-2-nitroethenyl phenylboronic acid, 4-e-2-nitroethenyl phenyl boronic acid
B(C1=CC=C(C=C1)C=C[N+](=O)[O-])(O)O
192.965
192.97
trans-4-(2-Nitrovinyl)benzeneboronic acid
1g
C8H8BNO4
3293756
GMEGTAODWQPCOD-AATRIKPKSA-N
[4-[(E)-2-nitroethenyl]phenyl]boronic acid
5708381
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning