missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3-Octene, 97%

$38.76 - $132.84

Chemical Identifiers

CAS 14919-01-8
Molecular Formula C8H16
Formula Weight 112.22
MDL Number MFCD00039971
UN Number UN3295
Chemical Name or Material trans-3-Octene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4315404 SDS Alfa Aesar™
4315404
2g Each for $38.76
AA4315409 SDS Alfa Aesar™
4315409
10g Each for $132.84
AA4315418 SDS Alfa Aesar™
4315418
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

14919-01-8
112.22
UN3295
C8H16
MFCD00039971
trans-3-Octene

Specifications

14919-01-8
Liquid
112.22
0.716g/mL
121° to 222°C
13°C (57°F)
trans-3-Octene
1.4130
C8H16
MFCD00039971
UN3295
2g
Characteristic
97%

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.