missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
$165.33 - $303.38
Chemical Identifiers
CAS | 14755-02-3 |
---|---|
Molecular Formula | C9H8O3 |
Molecular Weight (g/mol) | 164.16 |
MDL Number | MFCD00004390 |
InChI Key | KKSDGJDHHZEWEP-SNAWJCMRSA-N |
Synonym | 3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid |
PubChem CID | 637541 |
ChEBI | CHEBI:32357 |
IUPAC Name | (E)-3-(3-hydroxyphenyl)prop-2-enoic acid |
SMILES | C1=CC(=CC(=C1)O)C=CC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1446618
|
Thermo Scientific Chemicals
A1446618 |
50 g |
Each for $165.33
|
|
|||||
AAA1446622
|
Thermo Scientific Chemicals
A1446622 |
100 g |
Each for $303.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14755-02-3 | |
164.16 | |
KKSDGJDHHZEWEP-SNAWJCMRSA-N | |
637541 | |
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid |
C9H8O3 | |
MFCD00004390 | |
3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid | |
CHEBI:32357 | |
C1=CC(=CC(=C1)O)C=CC(=O)O |
Specifications
14755-02-3 | |
C9H8O3 | |
50 g | |
3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid | |
C1=CC(=CC(=C1)O)C=CC(=O)O | |
164.16 | |
CHEBI:32357 | |
99% |
192°C to 196°C | |
MFCD00004390 | |
2690254 | |
KKSDGJDHHZEWEP-SNAWJCMRSA-N | |
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid | |
637541 | |
164.16 | |
trans-3-Hydroxycinnamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-615-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only