missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-3-Hexenoic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1342818

 View more versions of this product

Catalog No. AAA1342818



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-3-Hexenoic acid
11°C to 12°C
206°C
Rancid
1.4398
50 g
1700862
XXHDAWYDNSXJQM-ONEGZZNKSA-N
(E)-hex-3-enoic acid
5282708
114.14
1577-18-0
0.961
99°C (210°F)
C6H10O2
MFCD00002786
UN3265
trans-3-hexenoic acid, hydrosorbic acid, 3-hexenoic acid, e-hex-3-enoic acid, 3-hexenoic acid, 3e, e-3-hexenoic acid, 3-hexenoic acid, e, trans-hex-3-enoic acid, unii-9b3n069edg, fema no. 3170
CC\C=C\CC(O)=O
114.14
CHEBI:49285
97%

Chemical Identifiers

1577-18-0
114.14
XXHDAWYDNSXJQM-ONEGZZNKSA-N
5282708
(E)-hex-3-enoic acid
C6H10O2
MFCD00002786
trans-3-hexenoic acid, hydrosorbic acid, 3-hexenoic acid, e-hex-3-enoic acid, 3-hexenoic acid, 3e, e-3-hexenoic acid, 3-hexenoic acid, e, trans-hex-3-enoic acid, unii-9b3n069edg, fema no. 3170
CHEBI:49285
CC\C=C\CC(O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 216-417-8

RTECSNumber : MP7730000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only