Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3-Hexene, 98%

Chemical Identifiers

Chemical Name or Material trans-3-Hexene
CAS 13269-52-8
Molecular Formula C6H12
Formula Weight 84.16
MDL Number MFCD00009386
UN Number UN3295
Beilstein 1718859
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1031103

 
alfa aesar™
L1031103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1031106

 
alfa aesar™
L1031106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 84.16
Chemical Name or Material trans-3-Hexene
Assay 98%
Molecular Formula C6H12
UN Number UN3295
Health Hazard 1 Danger
Odor Mild
Health Hazard 2 GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
MDL Number MFCD00009386
Beilstein 1718859
Melting Point −113°C
Boiling Point 66° to 67°C
Quantity 1g
Flash Point −25°C (−13°F)
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 13269-52-8

Applications
Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.

Solubility
Insoluble in water. Soluble in benzene, ligroin, ethanol, chloroform, ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.