missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3-Heptene, 97%

Manufacturer:  Alfa Aesar™ H5348214

 View more versions of this product

Catalog No. AAH5348214



Specifications

Specifications

trans-3-Heptene
−1°C (30°F)
94°C
25g
C7H14
UN3295
WZHKDGJSXCTSCK-FNORWQNLSA-N
(E)-hept-3-ene
5357259
97%
0.698
1.403
-137°C
14686-14-7
MFCD00009475
3-heptene c,t
CCCC=CCC
98.189
98.19

Chemical Identifiers

14686-14-7
98.189
WZHKDGJSXCTSCK-FNORWQNLSA-N
5357259
CCCC=CCC
C7H14
MFCD00009475
3-heptene c,t
(E)-hept-3-ene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 238-728-8

TSCA : No

Recommended Storage : Ambient temperatures

Documents