Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3-Ethoxycinnamic acid, 99%

$397.47 - $397.47

Chemical Identifiers

Chemical Name or Material trans-3-Ethoxycinnamic acid
CAS 103986-73-8
Molecular Formula C11H12O3
Formula Weight 192.22g/mol
MDL Number MFCD00016843
Beilstein 3199271
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0860714

 
alfa aesar™
L08607-14
25g Each for $397.47
Description & Specifications

Specifications

Quantity 25g
Melting Point 132° to 134°C
Quantity 25g
Chemical Name or Material trans-3-Ethoxycinnamic acid
CAS 103986-73-8
Assay 99%
Molecular Formula C11H12O3
Formula Weight 192.22g/mol
MDL Number MFCD00016843
Beilstein 3199271
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.