Please login to your online account to display your discounted pricing

trans-3-chloroacrylic acid, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC108550050

 View more versions of this product

Catalog No. AC108550050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Flakes
Assay ≥98.5%
Formula Weight 106.51
Quantity 5g
Color Light gray to beige-gray
Melting Point 82° to 85°C
Beilstein 02, 400
Chemical Name or Material trans-3-chloroacrylic acid
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C3H3ClO2
MDL Number MFCD00064237
CAS 2345-61-1
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Linear Formula ClCH=CHCO2H

Specifications

Chemical Name or Material trans-3-chloroacrylic acid
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3H3ClO2
Linear Formula ClCH=CHCO2H
Formula Weight 106.51
MDL Number MFCD00064237
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents