missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3-Chloro-beta-nitrostyrene, 97%

Manufacturer:  Alfa Aesar™ H6405306

 View more versions of this product

Catalog No. AAH6405306



Specifications

Specifications

trans-3-Chloro-beta-nitrostyrene
48°C to 52°C
37888-03-2
MFCD00175518
1-chloro-3-2-nitrovinyl benzene, a-nitrostyrene, e-1-chloro-3-2-nitrovinyl benzene, 1-3-chlorophenyl-2-nitroethene, trans-3-chloro-beta-nitrostyrene, beta-nitro-3-chlorostyrene, trans-1-chloro-3-2-nitro-vinyl-benzene, 1-chloro-3-e-2-nitrovinyl benzene, trans-1-3-chlorophenyl-2-nitroethene, cyto9f2, trans-3-chloro-
C1=CC(=CC(=C1)Cl)C=C[N+](=O)[O-]
183.591
183.59
97%
Yellow
5g
C8H6ClNO2
2087804
GXQRAWTWDNHGBS-SNAWJCMRSA-N
1-chloro-3-[(E)-2-nitroethenyl]benzene
5702314
Powder

Chemical Identifiers

37888-03-2
183.591
GXQRAWTWDNHGBS-SNAWJCMRSA-N
5702314
C1=CC(=CC(=C1)Cl)C=C[N+](=O)[O-]
C8H6ClNO2
MFCD00175518
1-chloro-3-2-nitrovinyl benzene, a-nitrostyrene, e-1-chloro-3-2-nitrovinyl benzene, 1-3-chlorophenyl-2-nitroethene, trans-3-chloro-beta-nitrostyrene, beta-nitro-3-chlorostyrene, trans-1-chloro-3-2-nitro-vinyl-benzene, 1-chloro-3-e-2-nitrovinyl benzene, trans-1-3-chlorophenyl-2-nitroethene, cyto9f2, trans-3-chloro-
1-chloro-3-[(E)-2-nitroethenyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents