Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3-Bromocinnamic acid, 98+%

$64.80 - $718.84

Chemical Identifiers

Chemical Name or Material trans-3-Bromocinnamic acid
CAS 14473-91-7
Molecular Formula C9H7BrO2
Formula Weight 227.06
MDL Number MFCD00004382
Beilstein 2084230
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1603506

 
alfa aesar™
A1603506
5g Each for $64.80

AAA1603514

 
alfa aesar™
A1603514
25g Each for $221.66

AAA1603522

 
alfa aesar™
A1603522
100g Each for $718.84
Description & Specifications

Specifications

Formula Weight 227.06
Chemical Name or Material trans-3-Bromocinnamic acid
TSCA TSCA
Assay ≥98%
Molecular Formula C9H7BrO2
MDL Number MFCD00004382
CAS 14473-91-7
Beilstein 2084230
Melting Point 178° to 182°C
Health Hazard 1 Warning
Quantity 5g
Odor Odorless
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GD8420000

Applications
trans-3-Bromocinnamic acid is used as organic synthesis of raw materials, fine chemicals, pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.