missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3-(Boc-amino)cyclohexanemethanol, 97%

$337.73 - $1,011.64

Chemical Identifiers

CAS 920966-17-2
Molecular Formula C12H23NO3
Molecular Weight (g/mol) 229.32
MDL Number MFCD03844607
InChI Key LBJSEPNOVVUVJA-NXEZZACHSA-N
Synonym tert-butyl trans-3-hydroxymethyl-cyclohexylcarbamate, tert-butyl trans-3-hydroxymethyl cyclohexylcarbamate, tert-butyl trans-3-hydroxymethylcyclohexyl carbamate, tert-butyl trans-3-hydroxymethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate
PubChem CID 24720931
IUPAC Name tert-butyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62934MD View Documents Alfa Aesar™
H62934MD
250mg Each for $337.73
 
AAH6293403 View Documents Alfa Aesar™
H6293403
1g Each for $1,011.64
 
Specifications

Chemical Identifiers

920966-17-2
229.32
LBJSEPNOVVUVJA-NXEZZACHSA-N
24720931
CC(C)(C)OC(=O)NC1CCCC(C1)CO
C12H23NO3
MFCD03844607
tert-butyl trans-3-hydroxymethyl-cyclohexylcarbamate, tert-butyl trans-3-hydroxymethyl cyclohexylcarbamate, tert-butyl trans-3-hydroxymethylcyclohexyl carbamate, tert-butyl trans-3-hydroxymethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate
tert-butyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate

Specifications

250mg
C12H23NO3
tert-butyl trans-3-hydroxymethyl-cyclohexylcarbamate, tert-butyl trans-3-hydroxymethyl cyclohexylcarbamate, tert-butyl trans-3-hydroxymethylcyclohexyl carbamate, tert-butyl trans-3-hydroxymethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate
CC(C)(C)OC(=O)NC1CCCC(C1)CO
229.32
229.31
trans-3-(Boc-amino)cyclohexanemethanol
920966-17-2
MFCD03844607
LBJSEPNOVVUVJA-NXEZZACHSA-N
tert-butyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate
24720931
97%