missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3-(Benzyloxymethyl)cyclobutanol, 97%

$211.27 - $632.27

Chemical Identifiers

CAS 172324-65-1
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD06657677
InChI Key KVQOEODTOJCIPA-UHFFFAOYSA-N
Synonym cis-3-benzyloxymethylcyclobutanol, trans-3-benzyloxymethylcyclobutanol, trans-3-benzyloxy methyl cyclobutanol, cis-3-benzyloxy methyl cyclobutanol, 1r,3r-3-benzyloxy methyl cyclobutanol, cyclobutanol,3-phenylmethoxy methyl-, cis, cis-3-benzyloxy methyl cyclobutanol, trans-3-benzyloxy methyl cyclobutanol, 3-benzyloxy methyl cyclobutan-1-ol, 1r,3r-3-benzyloxy methyl cyclobutan-1-ol
PubChem CID 10442570
IUPAC Name 3-(phenylmethoxymethyl)cyclobutan-1-ol
SMILES C1C(CC1O)COCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62088MD View Documents Alfa Aesar™
H62088MD
250mg Each for $211.27
 
AAH6208803 View Documents Alfa Aesar™
H6208803
1g Each for $632.27
 
Description

Description

Applications
trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

172324-65-1
192.258
KVQOEODTOJCIPA-UHFFFAOYSA-N
10442570
C1C(CC1O)COCC2=CC=CC=C2
C12H16O2
MFCD06657677
cis-3-benzyloxymethylcyclobutanol, trans-3-benzyloxymethylcyclobutanol, trans-3-benzyloxy methyl cyclobutanol, cis-3-benzyloxy methyl cyclobutanol, 1r,3r-3-benzyloxy methyl cyclobutanol, cyclobutanol,3-phenylmethoxy methyl-, cis, cis-3-benzyloxy methyl cyclobutanol, trans-3-benzyloxy methyl cyclobutanol, 3-benzyloxy methyl cyclobutan-1-ol, 1r,3r-3-benzyloxy methyl cyclobutan-1-ol
3-(phenylmethoxymethyl)cyclobutan-1-ol

Specifications

250mg
C12H16O2
cis-3-benzyloxymethylcyclobutanol, trans-3-benzyloxymethylcyclobutanol, trans-3-benzyloxy methyl cyclobutanol, cis-3-benzyloxy methyl cyclobutanol, 1r,3r-3-benzyloxy methyl cyclobutanol, cyclobutanol,3-phenylmethoxy methyl-, cis, cis-3-benzyloxy methyl cyclobutanol, trans-3-benzyloxy methyl cyclobutanol, 3-benzyloxy methyl cyclobutan-1-ol, 1r,3r-3-benzyloxy methyl cyclobutan-1-ol
C1C(CC1O)COCC2=CC=CC=C2
192.258
192.25
trans-3-(Benzyloxymethyl)cyclobutanol
172324-65-1
MFCD06657677
KVQOEODTOJCIPA-UHFFFAOYSA-N
3-(phenylmethoxymethyl)cyclobutan-1-ol
10442570
97%