missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-3-(Benzyloxymethyl)cyclobutanol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H62088MD

 View more versions of this product

Catalog No. AAH62088MD



Description

Description

trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

trans-3-(Benzyloxymethyl)cyclobutanol
C12H16O2
250 mg
Slightly soluble in water.
OC1CC(COCC2=CC=CC=C2)C1
192.26
192.25
172324-65-1
MFCD06657676,MFCD06657677
cis-3-benzyloxymethylcyclobutanol, trans-3-benzyloxymethylcyclobutanol, trans-3-benzyloxy methyl cyclobutanol, cis-3-benzyloxy methyl cyclobutanol, 1r,3r-3-benzyloxy methyl cyclobutanol, cyclobutanol,3-phenylmethoxy methyl-, cis, 3-benzyloxy methyl cyclobutan-1-ol, 1r,3r-3-benzyloxy methyl cyclobutan-1-ol
KVQOEODTOJCIPA-UHFFFAOYSA-N
3-(phenylmethoxymethyl)cyclobutan-1-ol
10442570
97%

Chemical Identifiers

172324-65-1
192.26
KVQOEODTOJCIPA-UHFFFAOYSA-N
10442570
OC1CC(COCC2=CC=CC=C2)C1
C12H16O2
MFCD06657676,MFCD06657677
cis-3-benzyloxymethylcyclobutanol, trans-3-benzyloxymethylcyclobutanol, trans-3-benzyloxy methyl cyclobutanol, cis-3-benzyloxy methyl cyclobutanol, 1r,3r-3-benzyloxy methyl cyclobutanol, cyclobutanol,3-phenylmethoxy methyl-, cis, 3-benzyloxy methyl cyclobutan-1-ol, 1r,3r-3-benzyloxy methyl cyclobutan-1-ol
3-(phenylmethoxymethyl)cyclobutan-1-ol
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only