missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3-Aminocyclohexanecarboxamide hydrochloride, 97%

$231.32 - $689.32

Chemical Identifiers

CAS 920966-29-6
Molecular Formula C7H15ClN2O
Molecular Weight (g/mol) 178.66
MDL Number MFCD12022628
InChI Key ZCFBGXVDLNNMDE-KGZKBUQUSA-N
Synonym trans-3-aminocyclohexanecarboxamide hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxamide hydrochloride
PubChem CID 46172883
IUPAC Name (1R,3R)-3-aminocyclohexane-1-carboxamide;hydrochloride
SMILES C1CC(CC(C1)N)C(=O)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62529MD View Documents Alfa Aesar™
H62529MD
250mg Each for $231.32
 
AAH6252903 View Documents Alfa Aesar™
H6252903
1g Each for $689.32
 
Specifications

Chemical Identifiers

920966-29-6
178.66
ZCFBGXVDLNNMDE-KGZKBUQUSA-N
46172883
C1CC(CC(C1)N)C(=O)N.Cl
C7H15ClN2O
MFCD12022628
trans-3-aminocyclohexanecarboxamide hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxamide hydrochloride
(1R,3R)-3-aminocyclohexane-1-carboxamide;hydrochloride

Specifications

250mg
C7H15ClN2O
trans-3-aminocyclohexanecarboxamide hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxamide hydrochloride
C1CC(CC(C1)N)C(=O)N.Cl
178.66
178.65
trans-3-Aminocyclohexanecarboxamide hydrochloride
920966-29-6
MFCD12022628
ZCFBGXVDLNNMDE-KGZKBUQUSA-N
(1R,3R)-3-aminocyclohexane-1-carboxamide;hydrochloride
46172883
97%