Please login to your online account to display your discounted pricing

trans-3,4-Difluorocinnamic acid, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC311580050

 View more versions of this product

Catalog No. AC311580050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Shiny Powder
Assay ≥96.0%
Formula Weight 184.14
Quantity 5g
Color Almost white
Melting Point 193° to 198°C
Chemical Name or Material trans-3, 4-Difluorocinnamic acid
Linear Formula FC6H3CH=CHCOH
CAS 112897-97-9
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
MDL Number MFCD00010320
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C9H6F2O2

Specifications

Chemical Name or Material trans-3, 4-Difluorocinnamic acid
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H6F2O2
Linear Formula FC6H3CH=CHCOH
Formula Weight 184.14
MDL Number MFCD00010320
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents