missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-3,4-Difluorocinnamic acid, 97%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C9H6F2O2
Linear Formula FC6H3CH=CHCOH
Formula Weight 184.14
MDL Number MFCD00010320
Chemical Name or Material trans-3, 4-Difluorocinnamic acid
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC311580050 SDS Thermo Fisher Scientific
AC311580050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC311580010 SDS Thermo Fisher Scientific
311580010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

97%
FC6H3CH=CHCOH
MFCD00010320
≥96.0%
C9H6F2O2
184.14
trans-3, 4-Difluorocinnamic acid

Specifications

Shiny Powder
5g
193° to 198°C
FC6H3CH=CHCOH
Authentic
C9H6F2O2
trans-3, 4-Difluorocinnamic acid
184.14
Almost white
112897-97-9
MFCD00010320
Glass bottle
97%
≥96.0%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.