missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl, 97%

Manufacturer:  Alfa Aesar™ H5535906

 View more versions of this product

Catalog No. AAH5535906



Description

Description

Solubility
Sparingly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl
5g
C21H24F2
1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
314.42
314.42
66°C to 68°C
85312-59-0
MFCD09838999
VULXHDGYVHCLLN-UHFFFAOYSA-N
1,2-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene
612306
97%

Chemical Identifiers

85312-59-0
314.42
VULXHDGYVHCLLN-UHFFFAOYSA-N
612306
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
C21H24F2
MFCD09838999
1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl
1,2-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents