missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl, 97%

Manufacturer:  Alfa Aesar™ H5535903

 View more versions of this product

Catalog No. AAH5535903



Description

Description

Solubility
Sparingly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl
1g
C21H24F2
1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl
VULXHDGYVHCLLN-UHFFFAOYSA-N
1,2-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene
612306
97%
66°C to 68°C
85312-59-0
MFCD09838999
Sparingly soluble in water.
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
314.42
314.42

Chemical Identifiers

85312-59-0
314.42
VULXHDGYVHCLLN-UHFFFAOYSA-N
612306
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
C21H24F2
MFCD09838999
1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl
1,2-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

Documents