missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3,4-Difluoro-4'-(4-n-pentylcyclohexyl)biphenyl, 97%

$92.26 - $366.45

Chemical Identifiers

CAS 134412-17-2
Molecular Formula C23H28F2
Molecular Weight (g/mol) 342.474
MDL Number MFCD09839000
InChI Key NQLHGECGZRJQLF-UHFFFAOYSA-N
Synonym 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl
PubChem CID 612307
IUPAC Name 1,2-difluoro-4-[4-(4-pentylcyclohexyl)phenyl]benzene
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5583503 View Documents Alfa Aesar™
H5583503
1g Each for $92.26
 
AAH5583506 View Documents Alfa Aesar™
H5583506
5g Each for $366.45
 
Description

Description

Applications
trans-3,4-Difluoro-4′-(4-n-pentylcyclohexyl)biphenyl, is used to make PDLC film, smart film, switchable film, PDLC glass, smart glass, switchable glass, PDLC mixture. It also acts as a liquid crystal monomer.

Solubility
Sparingly Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

134412-17-2
342.474
NQLHGECGZRJQLF-UHFFFAOYSA-N
612307
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
C23H28F2
MFCD09839000
1,1'-biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl
1,2-difluoro-4-[4-(4-pentylcyclohexyl)phenyl]benzene

Specifications

56°C to 57°C
134412-17-2
MFCD09839000
Sparingly Soluble in water.
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
342.474
342.47
trans-3,4-Difluoro-4'-(4-n-pentylcyclohexyl)biphenyl
1g
C23H28F2
1,1'-biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl
NQLHGECGZRJQLF-UHFFFAOYSA-N
1,2-difluoro-4-[4-(4-pentylcyclohexyl)phenyl]benzene
612307
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures