missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-3,4-Difluoro-4'-(4-n-pentylcyclohexyl)biphenyl, 97%

Manufacturer:  Alfa Aesar™ H5583506

 View more versions of this product

Catalog No. AAH5583506



Description

Description

Applications
trans-3,4-Difluoro-4′-(4-n-pentylcyclohexyl)biphenyl, is used to make PDLC film, smart film, switchable film, PDLC glass, smart glass, switchable glass, PDLC mixture. It also acts as a liquid crystal monomer.

Solubility
Sparingly Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

trans-3,4-Difluoro-4'-(4-n-pentylcyclohexyl)biphenyl
5g
C23H28F2
1,1'-biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
342.474
342.47
56°C to 57°C
134412-17-2
MFCD09839000
NQLHGECGZRJQLF-UHFFFAOYSA-N
1,2-difluoro-4-[4-(4-pentylcyclohexyl)phenyl]benzene
612307
97%

Chemical Identifiers

134412-17-2
342.474
NQLHGECGZRJQLF-UHFFFAOYSA-N
612307
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
C23H28F2
MFCD09839000
1,1'-biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl
1,2-difluoro-4-[4-(4-pentylcyclohexyl)phenyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents