Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2-Undecenoic acid, tech. 80%

$49.11 - $49.11

Chemical Identifiers

Chemical Name or Material trans-2-Undecenoic acid
CAS 15790-94-0
Purity Grade Technical Grade
Molecular Formula C11H20O2
Formula Weight 184.28
MDL Number MFCD00040909
Beilstein 1722519
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1157903

  SDS
alfa aesar™
L1157903
1g Each for $49.11
Description & Specifications

Specifications

Formula Weight 184.28
CAS 15790-94-0
Chemical Name or Material trans-2-Undecenoic acid
MDL Number MFCD00040909
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Beilstein 1722519
Molecular Formula C11H20O2
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 138° to 140°C (2mmHg)
Quantity 1g
Assay 80%
Purity Grade Technical Grade
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.