missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2-Undecenoic acid, tech. 80%

Manufacturer:  Alfa Aesar™ L1157903

 View more versions of this product

Catalog No. AAL1157903



Specifications

Specifications

trans-2-Undecenoic acid
1g
C11H20O2
1722519
IGBBVTAVILYDIO-MDZDMXLPSA-N
(E)-undec-2-enoic acid
5282728
184.28
Technical
138°C to 140°C (2mmHg)
15790-94-0
MFCD00040909
2-undecenoic acid, e-undec-2-enoic acid, trans-2-undecenoic acid, 2-hendecenoic acid, e-2-undecenoic acid, 2e-2-undecenoic acid, 2-undecensaeure, 2e-undecylenate, 2-undecensaeuren, undec-2-ensaeure
CCCCCCCCC=CC(=O)O
184.279
CHEBI:39450
80%

Chemical Identifiers

15790-94-0
184.279
IGBBVTAVILYDIO-MDZDMXLPSA-N
5282728
(E)-undec-2-enoic acid
C11H20O2
MFCD00040909
2-undecenoic acid, e-undec-2-enoic acid, trans-2-undecenoic acid, 2-hendecenoic acid, e-2-undecenoic acid, 2e-2-undecenoic acid, 2-undecensaeure, 2e-undecylenate, 2-undecensaeuren, undec-2-ensaeure
CHEBI:39450
CCCCCCCCC=CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents