Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2-Undecenal, 95%, remainder mainly cis-isomer

$192.41 - $192.41

Chemical Identifiers

Chemical Name or Material trans-2-Undecenal
CAS 53448-07-0
Molecular Formula C11H20O
Formula Weight 168.28
MDL Number MFCD00014680
Beilstein 1762239
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0643714

 
alfa aesar™
L0643714
25g Each for $192.41
Description & Specifications

Specifications

Formula Weight 168.28
Assay Percent Notes remainder mainly cis-isomer
CAS 53448-07-0
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C11H20O
MDL Number MFCD00014680
Health Hazard 1 Warning
Chemical Name or Material trans-2-Undecenal
Assay 95%
Beilstein 1762239
Boiling Point 115°C (10mmHg)
Quantity 25g
Flash Point 104°C (219°F)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C