Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2-Undecen-1-ol, 96%

$115.12 - $439.97

Chemical Identifiers

Chemical Name or Material trans-2-Undecen-1-ol
CAS 75039-84-8
Molecular Formula C11H22O
Formula Weight 170.3
MDL Number MFCD00014060
Beilstein 1841046
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1969806

  SDS
alfa aesar™
A1969806
5g Each for $115.12

AAA1969814

  SDS
alfa aesar™
A1969814
25g Each for $439.97
Description & Specifications

Specifications

Formula Weight 170.3
Flash Point >110°C (230°F)
TSCA TSCA
MDL Number MFCD00014060
Boiling Point 150°C (5mmHg)
Molecular Formula C11H22O
CAS 75039-84-8
Chemical Name or Material trans-2-Undecen-1-ol
Assay 96%
Quantity 5g
Beilstein 1841046

Applications
trans-2-Undecen-1-ol is used as perfuming agents.

Solubility
Soluble in water (67.15g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.