Please login to your online account to display your discounted pricing

trans-2-Phenylcyclopropane-1-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material trans-2-Phenylcyclopropane-1-carboxylic acid
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C10H10O2
Linear Formula C6H5C3H4CO2H
Formula Weight 162.19
MDL Number MFCD00001292
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC130460250

  SDS
thermo fisher scientific
AC130460250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 162.19
Assay ≥96.0% (GC)
Quantity 25g
Color Yellow or beige
Melting Point 88° to 92°C
Linear Formula C6H5C3H4CO2H
Molecular Formula C10H10O2
CAS 939-90-2
Assay Percent Range 97%
Infrared Spectrum Authentic
MDL Number MFCD00001292
Chemical Name or Material trans-2-Phenylcyclopropane-1-carboxylic acid
Packaging Glass bottle