Please login to your online account to display your discounted pricing

trans-2-Phenylcyclopropane-1-carboxylic acid, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC130460250

 View more versions of this product

Catalog No. AC130460250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 162.19
Assay ≥96.0% (GC)
Quantity 25g
Color Yellow or beige
Melting Point 88° to 92°C
Linear Formula C6H5C3H4CO2H
Molecular Formula C10H10O2
CAS 939-90-2
Assay Percent Range 97%
Infrared Spectrum Authentic
MDL Number MFCD00001292
Chemical Name or Material trans-2-Phenylcyclopropane-1-carboxylic acid
Packaging Glass bottle

Specifications

Chemical Name or Material trans-2-Phenylcyclopropane-1-carboxylic acid
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C10H10O2
Linear Formula C6H5C3H4CO2H
Formula Weight 162.19
MDL Number MFCD00001292