missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Pentene 95%, ACROS Organics™

$182.40 - $182.40

Chemical Identifiers

CAS 646-04-8
Assay Percent Range 95%
Molecular Formula C5H10
Linear Formula C2H5CH=CHCH3
Formula Weight 70.13
MDL Number MFCD00009384
Chemical Name or Material trans-2-Pentene, 95%
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC150000050 SDS Thermo Fisher Scientific
150000050
Glass bottle 5g Each for $182.40
Specifications Description & Specifications

Chemical Identifiers

646-04-8
C5H10
70.13
trans-2-Pentene, 95%
95%
C2H5CH=CHCH3
MFCD00009384
≥94%

Specifications

0.6400g/mL
Authentic
1.3785 to 1.3805 (20°C, 589nm)
Liquid
C5H10
C2H5CH=CHCH3
MFCD00009384
15, 7235
≥94%
646-04-8
Glass bottle
Clear colorless to slightly yellow
95%
5g
70.13
01, 210
trans-2-Pentene, 95%

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-461-4

RTECSNumber : SB2184500

TSCA : TSCA