Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2-Octenoic acid, 94%


Quantity 10g

Manufacturer: alfa aesar™  L0205809

 View more versions of this product

Catalog No. AAL0205809

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 142.2
MDL Number MFCD00002706
Molecular Formula C8H14O2
Assay 94%
Chemical Name or Material trans-2-Octenoic acid
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 1721115
Boiling Point 101° to 102°C (5mmHg)
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flash Point 140°C (284°F)
UN Number UN3265
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 10g
Odor Characteristic
CAS 1871-67-6

Specifications

Chemical Name or Material trans-2-Octenoic acid
CAS 1871-67-6
Molecular Formula C8H14O2
Formula Weight 142.2
MDL Number MFCD00002706
UN Number UN3265
Beilstein 1721115
Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents