missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2-Octenoic acid, 94%

Manufacturer:  Alfa Aesar™ L0205818

 View more versions of this product

Catalog No. AAL0205818



Specifications

Specifications

trans-2-Octenoic acid
Characteristic
50g
C8H14O2
UN3265
2-octenoic acid
CCCCCC=CC(=O)O
142.198
CHEBI:86544
94%
140°C (284°F)
101°C to 102°C (5mmHg)
1871-67-6
MFCD00002706
1721115
CWMPPVPFLSZGCY-VOTSOKGWSA-N
(E)-oct-2-enoic acid
5282714
142.2

Chemical Identifiers

1871-67-6
142.198
CWMPPVPFLSZGCY-VOTSOKGWSA-N
5282714
(E)-oct-2-enoic acid
C8H14O2
MFCD00002706
2-octenoic acid
CHEBI:86544
CCCCCC=CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents