Please login to your online account to display your discounted pricing

trans-2-Octene 98%, ACROS Organics™

$160.42 - $160.42

Chemical Identifiers

Chemical Name or Material trans-2-Octene
Name Note 98%
CAS 13389-42-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H16
Linear Formula CH3(CH2)4CH=CHCH3
Formula Weight 112.21
MDL Number MFCD00009532
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC149990100

  SDS
thermo fisher scientific
149990100
Glass bottle 10g Each for $160.42
Description & Specifications

Specifications

Formula Weight 112.21
Physical Form Liquid
Assay ≥97.5%
Quantity 10g
Color Clear colorless
EINECS Number 236-463-2
Density 0.7180g/mL
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
NMR Complies
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Refractive Index 1.4120 to 1.4140 (20°C, 589nm)
Infrared Spectrum Authentic
Molecular Formula C8H16
CAS 13389-42-9
Chemical Name or Material trans-2-Octene
Assay Percent Range 98%
MDL Number MFCD00009532
Name Note 98%
Linear Formula CH3(CH2)4CH=CHCH3
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.