missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Nitrocinnamic Acid, TCI America™

$192.50 - $375.84

Chemical Identifiers

CAS 1013-96-3
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00007189
InChI Key BBQDLDVSEDAYAA-AATRIKPKSA-N
Synonym 2-nitrocinnamic acid, o-nitrocinnamic acid, 3-2-nitrophenyl acrylic acid, e-3-2-nitrophenyl acrylic acid, cinnamic acid, o-nitro, trans-2-nitrocinnamic acid, 2e-3-2-nitrophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-nitrophenyl, 3-2-nitrophenyl-acrylic acid, ccris 1668
PubChem CID 735923
SMILES C1=CC=C(C(=C1)C=CC(=O)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N035610G View Documents TCI America
N035610G
10g Each for $192.50
 
N035625G View Documents TCI America
N035625G
25g Each for $375.84
 
Specifications

Chemical Identifiers

1013-96-3
193.158
BBQDLDVSEDAYAA-AATRIKPKSA-N
735923
C9H7NO4
MFCD00007189
2-nitrocinnamic acid, o-nitrocinnamic acid, 3-2-nitrophenyl acrylic acid, e-3-2-nitrophenyl acrylic acid, cinnamic acid, o-nitro, trans-2-nitrocinnamic acid, 2e-3-2-nitrophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-nitrophenyl, 3-2-nitrophenyl-acrylic acid, ccris 1668
C1=CC=C(C(=C1)C=CC(=O)O)[N+](=O)[O-]

Specifications

Yellow
1013-96-3
MFCD00007189
BBQDLDVSEDAYAA-AATRIKPKSA-N
193.158
193.16
≥98.0% (HPLC,T)
10g
C9H7NO4
2-nitrocinnamic acid, o-nitrocinnamic acid, 3-2-nitrophenyl acrylic acid, e-3-2-nitrophenyl acrylic acid, cinnamic acid, o-nitro, trans-2-nitrocinnamic acid, 2e-3-2-nitrophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-nitrophenyl, 3-2-nitrophenyl-acrylic acid, ccris 1668
C1=CC=C(C(=C1)C=CC(=O)O)[N+](=O)[O-]
735923
Crystal-Powder at 20°C
trans-2-Nitrocinnamic Acid
Safety and Handling

Safety and Handling

RTECSNumber : UD3642850

TSCA : No