missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Methyl-2-pentenoic Acid, TCI America™

$50.42 - $149.72

Chemical Identifiers

CAS 16957-70-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00002655
InChI Key JJYWRQLLQAKNAD-SNAWJCMRSA-N
Synonym 2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
PubChem CID 5365909
IUPAC Name (E)-2-methylpent-2-enoic acid
SMILES CCC=C(C)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28145G View Documents TCI America
M28145G
5g Each for $50.42
 
M281425G View Documents TCI America
M281425G
25g Each for $149.72
 
Specifications

Chemical Identifiers

16957-70-3
114.144
JJYWRQLLQAKNAD-SNAWJCMRSA-N
5365909
CCC=C(C)C(=O)O
C6H10O2
MFCD00002655
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
(E)-2-methylpent-2-enoic acid

Specifications

125°C
5g
C6H10O2
3265
JJYWRQLLQAKNAD-SNAWJCMRSA-N
(E)-2-methylpent-2-enoic acid
5365909
Clear Liquid at 20°C
trans-2-Methyl-2-pentenoic Acid
Yellow
16957-70-3
MFCD00002655
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
CCC=C(C)C(=O)O
114.144
114.14
≥97.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0609

TSCA : Yes