missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Methyl-2-pentenal, TCI America™

Manufacturer:  TCI America M089325ML

 View more versions of this product

Catalog No. M089325ML



Specifications

Specifications

trans-2-Methyl-2-pentenal
Yellow
14250-96-5
MFCD00006978
IDEYZABHVQLHAF-GQCTYLIASA-N
(E)-2-methylpent-2-enal
5319754
Clear Liquid at 20°C
138°C
25mL
C6H10O
1989
CCC=C(C)C=O
98.145
98.15
≥97.0% (GC)

Chemical Identifiers

14250-96-5
98.145
IDEYZABHVQLHAF-GQCTYLIASA-N
(E)-2-methylpent-2-enal
C6H10O
MFCD00006978
5319754
CCC=C(C)C=O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0520

TSCA : Yes

Documents