Learn More
trans-2-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
$80.62 - $228.48
Chemical Identifiers
CAS | 497-03-0 |
---|---|
Molecular Formula | C5H8O |
Molecular Weight (g/mol) | 84.118 |
MDL Number | MFCD00006977 |
InChI Key | ACWQBUSCFPJUPN-HWKANZROSA-N |
Synonym | tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e |
PubChem CID | 5321950 |
IUPAC Name | (E)-2-methylbut-2-enal |
SMILES | CC=C(C)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0811306
|
Thermo Scientific Chemicals
L0811306 |
5 g |
Each for $80.62
|
|
|||||
AAL0811314
|
Thermo Scientific Chemicals
L0811314 |
25 g |
Each for $228.48
|
|
|||||
Description
Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.
Solubility
Soluble in water
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
497-03-0 | |
84.118 | |
ACWQBUSCFPJUPN-HWKANZROSA-N | |
5321950 | |
CC=C(C)C=O |
C5H8O | |
MFCD00006977 | |
tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e | |
(E)-2-methylbut-2-enal |
Specifications
497-03-0 | |
117°C to 119°C | |
Fruit-like | |
1.448 | |
5 g | |
1698207 | |
tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e | |
ACWQBUSCFPJUPN-HWKANZROSA-N | |
(E)-2-methylbut-2-enal | |
5321950 | |
97% |
0.871 | |
18°C (64°F) | |
C5H8O | |
MFCD00006977 | |
UN1989 | |
Air sensitive | |
Soluble in water | |
CC=C(C)C=O | |
84.118 | |
84.12 | |
trans-2-Methyl-2-butenal |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber : 207-833-0
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only