missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 265872500

 View more versions of this product

Catalog No. AC265872500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride
Glass bottle
150°C to 155°C
99+%
99% min. (Argentometry)
MFCD00145424
Solubility in water: soluble. Other solubilities: soluble in methanol
C1CCC(C(C1)CO)N.Cl
165.66
165.66
99+%
Authentic
White
250mg
28250-45-5
C7H15NO·HCl
1r,2r-2-aminocyclohexyl methan-1-ol, chloride, 1r,2r-2-aminocyclohexyl methanol hydrochloride, cyclohexanemethanol, 2-amino-, hydrochloride, 1r,2r-rel, trans-+/--2-aminocyclohexyl methanol hydrochloride, trans-+/--2-aminocyclohexyl methanol hydrochloride salt, trans-2-amino-cyclohexyl-methanol hydrochloride, trans-2-aminocyclohexyl methanol hydrochloride, trans-2-aminocyclohexylmethanol hydrochloride, trans-2-hydroxymethyl-1-cyclohexylamine hydrochloride
ORPKUPNVXFLMRG-UOERWJHTSA-N
[(1R,2R)-2-aminocyclohexyl]methanol;hydrochloride
2724654
Crystalline Powder

Chemical Identifiers

28250-45-5
165.66
ORPKUPNVXFLMRG-UOERWJHTSA-N
2724654
C1CCC(C(C1)CO)N.Cl
C7H15NO·HCl
MFCD00145424
1r,2r-2-aminocyclohexyl methan-1-ol, chloride, 1r,2r-2-aminocyclohexyl methanol hydrochloride, cyclohexanemethanol, 2-amino-, hydrochloride, 1r,2r-rel, trans-+/--2-aminocyclohexyl methanol hydrochloride, trans-+/--2-aminocyclohexyl methanol hydrochloride salt, trans-2-amino-cyclohexyl-methanol hydrochloride, trans-2-aminocyclohexyl methanol hydrochloride, trans-2-aminocyclohexylmethanol hydrochloride, trans-2-hydroxymethyl-1-cyclohexylamine hydrochloride
[(1R,2R)-2-aminocyclohexyl]methanol;hydrochloride
SDS Documents