Please login to your online account to display your discounted pricing

trans-2-Hydroxy-1-cyclohexanecarboxamide, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material trans-2-Hydroxy-1-cyclohexanecarboxamide
Name Note 99%
CAS 24947-95-3
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H13NO2
Formula Weight 143.19
MDL Number MFCD00145423
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC277725000

  SDS
thermo fisher scientific
AC277725000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 143.19
Quantity 500mg
Color White
Melting Point 104° to 107°C
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material trans-2-Hydroxy-1-cyclohexanecarboxamide
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H13NO2
MDL Number MFCD00145423
CAS 24947-95-3
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.