missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Hexenyl acetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1878618

 View more versions of this product

Catalog No. AAA1878618



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-2-Hexenyl acetate
0.9
58°C (136°F)
1.427
50 g
1721851
HRHOWZHRCRZVCU-AATRIKPKSA-N
[(E)-hex-2-enyl] acetate
2733294
98%
2497-18-9
165°C to 166°C
C8H14O2
MFCD00009474
UN3272
z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z
CCCC=CCOC(=O)C
142.198
142.2

Chemical Identifiers

2497-18-9
142.198
HRHOWZHRCRZVCU-AATRIKPKSA-N
2733294
CCCC=CCOC(=O)C
C8H14O2
MFCD00009474
z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z
[(E)-hex-2-enyl] acetate
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 219-680-7

RTECSNumber : MP8425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only