missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Hexenal, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 158130250

 View more versions of this product

Catalog No. AC158130250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-2-Hexenal
6728-26-3
Colorless to Yellow
146.0°C to 149.0°C
Authentic
Glass bottle
1.4440 to 1.4500
MFCD00007008
01, I, 382
trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e
MBDOYVRWFFCFHM-SNAWJCMRSA-N
(E)-hex-2-enal
5281168
98.14
Liquid
99%
5mg KOH/g max.
0.8460g/mL
35°C
98.5% min. (GC)
C6H10O
CH3CH2CH2CH=CHCHO
25 mL
0.846
Solubility in water: insoluble.
CCCC=CC=O
98.14
CHEBI:28913
99%

Chemical Identifiers

6728-26-3
98.14
MBDOYVRWFFCFHM-SNAWJCMRSA-N
5281168
(E)-hex-2-enal
C6H10O
MFCD00007008
trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e
CHEBI:28913
CCCC=CC=O
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

EINECSNumber : 229-778-1

RTECSNumber : MP5900000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only