missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2-Hexenal, 96%

Manufacturer:  Alfa Aesar™ A1151322

 View more versions of this product

Catalog No. AAA1151322



Description

Description

Applications
trans-2-Hexenal is used in evaluation of quality of protected designation of origin virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. It was used in the multicomponent method for the determination of low-molecular-weight carbonyl compounds in biological samples.

Solubility
Not miscible in water.

Notes
Store at 4°C. Incompatible materials are bases, air, oxidizing agents, reducing agents and heat.
Specifications

Specifications

trans-2-Hexenal
Sweet
100g
C6H10O
UN1988
trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e
CCCC=CC=O
98.145
CHEBI:28913
96%
38°C (100°F)
37°C to 38°C (8mmHg)
6728-26-3
MFCD00007008
1699684
MBDOYVRWFFCFHM-SNAWJCMRSA-N
(E)-hex-2-enal
5281168
98.15

Chemical Identifiers

6728-26-3
98.145
MBDOYVRWFFCFHM-SNAWJCMRSA-N
5281168
(E)-hex-2-enal
C6H10O
MFCD00007008
trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e
CHEBI:28913
CCCC=CC=O
Safety and Handling

Safety and Handling

GHS H Statement
H311-H226-H302-H332-H315-H319-H335
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, FLAMMABLE, TOXIC, N.O.S.

RTECSNumber : MP5900000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents