Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2-Fluorocyclohexanol, 97%

$80.04 - $207.80

Chemical Identifiers

Chemical Name or Material trans-2-Fluorocyclohexanol
CAS 656-60-0
Molecular Formula C6H11FO
Formula Weight 118.15
MDL Number MFCD08146648
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2603103

  SDS
alfa aesar™
H2603103
1g Each for $207.80

AAH26031MD

  SDS
alfa aesar™
H26031MD
250mg Each for $80.04
Description & Specifications

Specifications

Formula Weight 118.15
Chemical Name or Material trans-2-Fluorocyclohexanol
Assay 97%
Molecular Formula C6H11FO
MDL Number MFCD08146648
CAS 656-60-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 166°C
Melting Point 21°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.