missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-2-Butenyl acetate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0973414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
trans-2-Butenyl acetate | |
0.919 | |
31°C (87°F) | |
1.42 | |
25 g | |
1720994 | |
WNHXJHGRIHUOTG-ONEGZZNKSA-N | |
[(E)-but-2-enyl] acetate | |
5363082 | |
95% |
628-08-0 | |
128°C to 130°C | |
C6H10O2 | |
MFCD00039912 | |
UN3272 | |
trans-2-butenyl acetate, crotyl acetate, e-2-butenyl acetate, 2-buten-1-ol, acetate, e-crotyl acetate, e-but-2-enyl acetate, 2-buten-1-ol, acetate, 2e, 2-buten-1-ol, 1-acetate, 2e, 2e-but-2-en-1-yl acetate, e-2-buten-1-yl acetate | |
CC=CCOC(=O)C | |
114.144 | |
114.15 |
Chemical Identifiers
628-08-0 | |
114.144 | |
WNHXJHGRIHUOTG-ONEGZZNKSA-N | |
5363082 | |
CC=CCOC(=O)C |
C6H10O2 | |
MFCD00039912 | |
trans-2-butenyl acetate, crotyl acetate, e-2-butenyl acetate, 2-buten-1-ol, acetate, e-crotyl acetate, e-but-2-enyl acetate, 2-buten-1-ol, acetate, 2e, 2-buten-1-ol, 1-acetate, 2e, 2e-but-2-en-1-yl acetate, e-2-buten-1-yl acetate | |
[(E)-but-2-enyl] acetate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only