Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2-Bromocinnamic acid, 98+%

$106.08 - $1,330.47

Chemical Identifiers

Chemical Name or Material trans-2-Bromocinnamic acid
CAS 7345-79-1
Molecular Formula C9H7BrO2
Formula Weight 227.06
MDL Number MFCD00016836
Beilstein 2357933
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1138106

  SDS
alfa aesar™
A1138106
5g Each for $106.08

AAA1138114

  SDS
alfa aesar™
A1138114
25g Each for $414.36

AAA1138122

  SDS
alfa aesar™
A1138122
100g Each for $1,330.47
Description & Specifications

Specifications

Formula Weight 227.06
Chemical Name or Material trans-2-Bromocinnamic acid
Molecular Formula C9H7BrO2
Beilstein 2357933
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Melting Point 216° to 219°C
CAS 7345-79-1
MDL Number MFCD00016836
Odor Odorless
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.