missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-2-(Boc-amino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H62007MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
trans-2-(Boc-amino)cyclohexanecarboxylic acid | |
C12H21NO4 | |
boc-1,2-trans-achc-oh, trans-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2r-boc-2-aminocyclohexane carboxylic acid, boc-trans-2-aminocyclohexanecarboxylic acid, trans-2-boc-amino cyclohexanecarboxylic acid, 1r,2r-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, trans-1-t-butyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, pubchem24192, trans-2-boc-amino-cyclohexanecarboxylic acid | |
CC(C)(C)OC(=O)NC1CCCCC1C(=O)O | |
243.303 | |
243.29 | |
250 mg |
209128-50-7 | |
MFCD02094138 | |
QJEQJDJFJWWURK-RKDXNWHRSA-N | |
(1R,2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid | |
1512488 | |
97% | |
Solid |
Chemical Identifiers
209128-50-7 | |
243.303 | |
QJEQJDJFJWWURK-RKDXNWHRSA-N | |
1512488 | |
CC(C)(C)OC(=O)NC1CCCCC1C(=O)O |
C12H21NO4 | |
MFCD02094138 | |
boc-1,2-trans-achc-oh, trans-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2r-boc-2-aminocyclohexane carboxylic acid, boc-trans-2-aminocyclohexanecarboxylic acid, trans-2-boc-amino cyclohexanecarboxylic acid, 1r,2r-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, trans-1-t-butyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, pubchem24192, trans-2-boc-amino-cyclohexanecarboxylic acid | |
(1R,2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only